siRNA Details
| Gene Symbol: Gm5136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203660.2 | NP_988991.1 | 1 | 608 | ||
| GenBank mRNA | AK134982.1 | - | 1 | 138 | ||
| BC120506.1 | - | 1 | 608 | 40129429 | ||
| BC120508.1 | - | 1 | 608 | 40129431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5136| Gene Symbol: Gm5136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203660.2 | NP_988991.1 | 1 | 608 | ||
| GenBank mRNA | AK134982.1 | - | 1 | 138 | ||
| BC120506.1 | - | 1 | 608 | 40129429 | ||
| BC120508.1 | - | 1 | 608 | 40129431 | ||