siRNA Details
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145059.1 | - | 3 | 303 | ||
BC010252.1 | - | 3 | 303 | |||
BC029177.1 | - | 3 | 281 | 3988386 | ||
U54563.1 | - | 3 | 286 |

More Information
Other Species Targeted

Panther Classification:
