siRNA Details
| Gene Symbol: Eif3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008388.2 | NP_032414.1 | 6 | 616 | ||
| GenBank mRNA | AK002576.1 | - | 6 | 593 | ||
| AK011436.1 | - | 6 | 586 | |||
| AK083874.1 | - | 1 | 2330 | |||
| AK145059.1 | - | 6 | 599 | |||
| AK146936.1 | - | 6 | 588 | |||
| AK161650.1 | - | 6 | 586 | |||
| AK165929.1 | - | 6 | 594 | |||
| BC010252.1 | - | 6 | 599 | |||
| BC029177.1 | - | 6 | 577 | 3988386 | ||

Eif3e
Genomic Map