siRNA Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008914.2 | NP_032940.1 | 14 | 2798 | ||
| GenBank mRNA | AK037615.1 | - | 1 | 1451 | ||
| BC037668.1 | - | 14 | 2621 | |||
| BC049860.1 | - | 1 | 1773 | |||
| BC066000.1 | - | 14 | 2770 | 5699179 | ||

Ppp3cb
Genomic Map