siRNA Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009402.2 | NP_033428.1 | 3 | 598 | ||
| GenBank mRNA | AF076482.1 | - | 3 | 597 | ||
| AF193843.1 | - | 3 | 586 | |||
| AK008335.1 | - | 3 | 599 | |||
| X86374.1 | - | 3 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pglyrp1| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009402.2 | NP_033428.1 | 3 | 598 | ||
| GenBank mRNA | AF076482.1 | - | 3 | 597 | ||
| AF193843.1 | - | 3 | 586 | |||
| AK008335.1 | - | 3 | 599 | |||
| X86374.1 | - | 3 | 595 | |||