siRNA Details
| Gene Symbol: Kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010615.1 | NP_034745.1 | 7 | 958 | ||
| GenBank mRNA | AB001427.1 | - | 4 | 470 | ||
| AJ223293.1 | - | 6 | 653 | |||
| AK083324.1 | - | 7 | 934 | |||
| BC060670.1 | - | 7 | 958 | 6406754 | ||

Kif11
Genomic Map