siRNA Details
| Gene Symbol: Pitpnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289472.1 | NP_001276401.1 | 3 | 526 | ||
| NM_011256.3 | NP_035386.1 | 4 | 717 | |||
| GenBank mRNA | AF058693.1 | - | 3 | 385 | ||
| AK015126.1 | - | 3 | 489 | |||
| AK129364.1 | - | 3 | 526 | |||
| AK147486.1 | - | 3 | 398 | |||
| BC070452.1 | - | 3 | 526 | 30621943 | ||
| BC082303.1 | - | 4 | 717 | |||

Pitpnm2
Genomic Map