siRNA Details
| Gene Symbol: Pitpnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289472.1 | NP_001276401.1 | 25 | 6462 | ||
| NM_011256.3 | NP_035386.1 | 25 | 6491 | |||
| GenBank mRNA | AF058693.1 | - | 24 | 6159 | ||
| BC070452.1 | - | 22 | 5751 | 30621943 | ||
| BC082303.1 | - | 25 | 6491 | |||

Pitpnm2
Genomic Map