siRNA Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011989.4 | NP_036119.1 | 2, 3 | 444 | ||
| GenBank mRNA | AJ251113.1 | - | 1, 2 | 157 | ||
| AK036919.1 | - | 2, 3 | 446 | |||
| AK155313.1 | - | 2, 3 | 414 | |||
| AK155388.1 | - | 2, 3 | 414 | |||
| AK170377.1 | - | 2, 3 | 448 | |||
| BC023114.1 | - | 2, 3 | 295 | 4158956 | ||
| CT010312.1 | - | 1, 2 | 157 | |||

Slc27a4
Genomic Map