siRNA Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011989.4 | NP_036119.1 | 3 | 814 | ||
| GenBank mRNA | AF072759.1 | - | 1 | 116 | ||
| AJ251113.1 | - | 2 | 527 | |||
| AK036919.1 | - | 3 | 816 | |||
| AK155313.1 | - | 3 | 783 | |||
| AK155388.1 | - | 3 | 784 | |||
| AK170377.1 | - | 3 | 818 | |||
| BC023114.1 | - | 3 | 665 | 4158956 | ||
| CT010312.1 | - | 2 | 527 | |||

Slc27a4
Genomic Map