siRNA Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013921.3 | NP_038949.2 | 6 | 1131 | ||
| GenBank mRNA | AJ243866.1 | - | 6 | 1094 | ||
| AK008263.1 | - | 5 | 976 | |||
| BC010970.1 | - | 6 | 1124 | 4216644 | ||
| BC040348.1 | - | 6 | 1131 | 4206523 | ||

Prss30
Genomic Map