siRNA Details
| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022322.2 | NP_071717.1 | 5 | 690 | ||
| GenBank mRNA | AB055422.1 | - | 5 | 497 | ||
| AF191768.1 | - | 5 | 636 | |||
| AF219993.1 | - | 5 | 563 | |||
| AF291655.1 | - | 5 | 715 | |||
| AK003748.1 | - | 5 | 574 | |||
| AK014761.1 | - | 5 | 529 | |||
| BC006919.1 | - | 5 | 499 | 3601223 | ||
| BC049944.1 | - | 5 | 690 | 2811628 | ||

Tnmd
Genomic Map