siRNA Details
| Gene Symbol: Gprasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004359.2 | NP_001004359.1 | 10 | 1625 | ||
| NM_001005385.1 | NP_001005385.1 | 10 | 1511 | |||
| NM_026081.5 | NP_080357.4 | 11 | 1714 | |||
| GenBank mRNA | AB093241.1 | - | 5 | 831 | ||
| AK050919.1 | - | 7 | 1149 | |||
| AK083192.1 | - | 10 | 1471 | |||
| BC058970.1 | - | 1 | 468 | 6826050 | ||
| BC070413.1 | - | 11 | 1662 | |||
| BC085157.1 | - | 10 | 1413 | 30357108 | ||

Gprasp1
Genomic Map