siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026112.4 | NP_080388.2 | 7 | 1052 | ||
| GenBank mRNA | AK010575.1 | - | 7 | 1047 | ||
| AK021179.1 | - | 7 | 1001 | |||
| AK044645.1 | - | 7 | 1020 | |||
| AK077352.1 | - | 3 | 476 | |||
| AK139269.1 | - | 7 | 1053 | |||
| AK143200.1 | - | 7 | 1017 | |||
| AK173278.1 | - | 8 | 1058 | |||
| BC052747.1 | - | 7 | 971 | 6837952 | ||

Zfp606
Genomic Map