siRNA Details
| Gene Symbol: Zfp606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026112.4 | NP_080388.2 | 7 | 1211 | ||
| GenBank mRNA | AK010575.1 | - | 7 | 1206 | ||
| AK021179.1 | - | 7 | 1160 | |||
| AK044645.1 | - | 7 | 1179 | |||
| AK077352.1 | - | 3 | 635 | |||
| AK139269.1 | - | 7 | 1212 | |||
| AK143200.1 | - | 7 | 1176 | |||
| AK173278.1 | - | 8 | 1217 | |||
| BC052747.1 | - | 7 | 1130 | 6837952 | ||

Zfp606
Genomic Map