siRNA Details
| Gene Symbol: Soga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026138.2 | NP_080414.2 | 4 | 1962 | ||
| GenBank mRNA | AK018139.1 | - | 3 | 492 | ||
| AK030682.1 | - | 4 | 1835 | |||
| AK163577.1 | - | 3 | 465 | |||
| BC066079.1 | - | 4 | 2119 | 30355520 | ||

Soga3
Genomic Map