siRNA Details
| Gene Symbol: Phf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290546.1 | NP_001277475.1 | 5 | 596 | ||
| NM_027642.2 | NP_081918.1 | 7 | 882 | |||
| GenBank mRNA | AK016480.1 | - | 7 | 883 | ||
| AK028498.1 | - | 5 | 518 | |||
| AK038320.1 | - | 7 | 802 | |||
| AK122560.1 | - | 7 | 812 | |||
| AK141804.1 | - | 7 | 825 | |||
| AK163345.1 | - | 7 | 836 | |||
| BC043127.1 | - | 7 | 830 | 6410361 | ||
| BC055330.1 | - | 7 | 825 | 6406483 | ||
| BC057374.1 | - | 7 | 788 | 6826091 | ||

Phf6
Genomic Map