siRNA Details
| Gene Symbol: Phf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290546.1 | NP_001277475.1 | 9 | 2207 | ||
| NM_027642.2 | NP_081918.1 | 11 | 2493 | |||
| GenBank mRNA | AK028498.1 | - | 9 | 2129 | ||
| AK087929.1 | - | 1 | 888 | |||
| AK122560.1 | - | 11 | 2423 | |||
| AK135407.1 | - | 3 | 1338 | |||
| AK141804.1 | - | 11 | 2437 | |||
| AK163345.1 | - | 11 | 2447 | |||
| BC043127.1 | - | 11 | 2441 | 6410361 | ||
| BC055330.1 | - | 11 | 2437 | 6406483 | ||
| BC057374.1 | - | 11 | 2399 | 6826091 | ||

Phf6
Genomic Map