siRNA Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079830.2 | NP_001073299.1 | 2 | 1249 | ||
| NM_053170.3 | NP_444400.2 | 2 | 1249 | |||
| GenBank mRNA | AK129293.1 | - | 2 | 508 | ||
| AY458590.1 | - | 2 | 1255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim33| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079830.2 | NP_001073299.1 | 2 | 1249 | ||
| NM_053170.3 | NP_444400.2 | 2 | 1249 | |||
| GenBank mRNA | AK129293.1 | - | 2 | 508 | ||
| AY458590.1 | - | 2 | 1255 | |||