siRNA Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323374.1 | NP_001310303.1 | 12 | 1637 | ||
| NM_001323375.1 | NP_001310304.1 | 12 | 1669 | |||
| NM_001323376.1 | NP_001310305.1 | 9 | 1639 | |||
| NM_001323377.1 | NP_001310306.1 | 10 | 1397 | |||
| NM_001744.5 | NP_001735.1 | 11 | 1785 | |||
| GenBank mRNA | AK311662.1 | - | 9 | 1494 | ||
| AL833368.1 | - | 2 | 1547 | |||
| BC016695.1 | - | 11 | 1629 | |||
| BC025687.1 | - | 11 | 1627 | 5205478 | ||
| D30742.1 | - | 11 | 1615 | |||

CAMK4
Genomic Map