siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290699.1 | NP_001277628.1 | 3 | 354 | ||
| NM_001290700.1 | NP_001277629.1 | 3 | 354 | |||
| NM_133817.3 | NP_598578.1 | 3 | 354 | |||
| GenBank mRNA | AK015797.1 | - | 3 | 329 | ||
| AK030088.1 | - | 3 | 187 | |||
| AK122541.1 | - | 3 | 179 | |||
| AK210231.1 | - | 2 | 101 | |||
| BC019546.1 | - | 3 | 190 | 4038791 | ||
| BC062154.1 | - | 3 | 144 | 6331597 | ||

Zfp451
Genomic Map