siRNA Details
| Gene Symbol: INHBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005538.3 | NP_005529.1 | 1 | 492 | ||
| GenBank mRNA | AK314849.1 | - | 1 | 464 | ||
| BC020693.1 | - | 1 | 434 | |||
| BC130324.1 | - | 1 | 373 | 40146355 | ||
| BC130326.1 | - | 1 | 373 | 40146357 | ||
| BP237373.1 | - | 1 | 492 | |||

INHBC
Genomic Map