siRNA Details
| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146191.3 | NP_666303.3 | 34 | 7195 | ||
| GenBank mRNA | AK076432.1 | - | 7 | 2522 | ||
| AK081849.1 | - | 4 | 2188 | |||
| BC066159.1 | - | 3 | 1717 | |||
| BC072664.1 | - | 24 | 5009 | 30543489 | ||

Lrrk1
Genomic Map