siRNA Details
| Gene Symbol: Zfp426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110309.1 | NP_001103779.1 | 9 | 768 | ||
| NM_146221.4 | NP_666333.1 | 7 | 906 | |||
| GenBank mRNA | AK036912.1 | - | 9 | 660 | ||
| AK085144.1 | - | 7 | 907 | |||
| AK220563.1 | - | 7 | 1608 | |||
| BC023931.1 | - | 9 | 770 | 5324400 | ||
| BC024805.1 | - | 9 | 764 | 5359610 | ||
| U29495.1 | - | 4 | 259 | |||

Zfp426
Genomic Map