siRNA Details
Gene Symbol: Olfr930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146272.1 | NP_666384.1 | 1 | 457 | ||
GenBank mRNA | BC119252.1 | - | 1 | 539 | 8734001 | |
BC119254.1 | - | 1 | 541 | 8734003 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146272.1 | NP_666384.1 | 1 | 457 | ||
GenBank mRNA | BC119252.1 | - | 1 | 539 | 8734001 | |
BC119254.1 | - | 1 | 541 | 8734003 |