siRNA Details
| Gene Symbol: Olfr1394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146276.1 | NP_666388.1 | 1 | 605 | ||
| GenBank mRNA | BC127968.1 | - | 1 | 609 | 40135034 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1394| Gene Symbol: Olfr1394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146276.1 | NP_666388.1 | 1 | 605 | ||
| GenBank mRNA | BC127968.1 | - | 1 | 609 | 40135034 | |