siRNA Details
| Gene Symbol: Olfr745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146299.2 | NP_666411.2 | 1 | 684 | ||
| GenBank mRNA | BC132579.1 | - | 1 | 682 | 40130856 | |
| BC132581.1 | - | 1 | 682 | 40130858 | ||
More Information
Set Membership: |
|

Olfr745
Genomic Map