siRNA Details
| Gene Symbol: Olfr420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146305.2 | NP_666417.2 | 1 | 580 | ||
| GenBank mRNA | BC145885.1 | - | 1 | 580 | 40131188 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr420| Gene Symbol: Olfr420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146305.2 | NP_666417.2 | 1 | 580 | ||
| GenBank mRNA | BC145885.1 | - | 1 | 580 | 40131188 | |