siRNA Details
| Gene Symbol: KLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002231.2 | NP_001002231.1 | 4 | 622 | ||
| NM_001256080.1 | NP_001243009.1 | 3 | 462 | |||
| NM_005551.4 | NP_005542.1 | 4 | 622 | |||
| NR_045762.1 | 4 | 609 | ||||
| NR_045763.1 | 4 | 708 | ||||
| GenBank mRNA | AF188745.1 | - | 4 | 584 | ||
| AF188746.1 | - | 4 | 597 | |||
| AF188747.1 | - | 4 | 597 | |||
| AK300527.1 | - | 4 | 708 | |||
| AK300549.1 | - | 3 | 456 | |||
| AK300566.1 | - | 4 | 616 | |||
| AK309968.1 | - | 2 | 584 | |||
| AK309969.1 | - | 4 | 548 | |||
| AK309973.1 | - | 3 | 443 | |||
| AK309982.1 | - | 2 | 575 | |||
| AK309985.1 | - | 3 | 459 | |||
| AK315890.1 | - | 3 | 436 | |||
| AY429509.1 | - | 3 | 723 | |||
| AY429510.1 | - | 2 | 563 | |||
| BC005196.1 | - | 4 | 622 | 3951036 | ||
| BT006650.1 | - | 4 | 581 | |||
| S39329.1 | - | 4 | 597 | |||

KLK2
Genomic Map