siRNA Details
| Gene Symbol: Olfr1309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146447.1 | NP_666658.1 | 1 | 819 | ||
| GenBank mRNA | BC128010.1 | - | 1 | 823 | 40136004 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1309| Gene Symbol: Olfr1309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146447.1 | NP_666658.1 | 1 | 819 | ||
| GenBank mRNA | BC128010.1 | - | 1 | 823 | 40136004 | |