siRNA Details
| Gene Symbol: Olfr282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146457.2 | NP_666668.2 | 1 | 474 | ||
| GenBank mRNA | BC128027.1 | - | 1 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr282| Gene Symbol: Olfr282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146457.2 | NP_666668.2 | 1 | 474 | ||
| GenBank mRNA | BC128027.1 | - | 1 | 477 | ||