siRNA Details
| Gene Symbol: Olfr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146488.1 | NP_666699.1 | 1 | 916 | ||
| GenBank mRNA | BC148231.1 | - | 1 | 920 | 40142242 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr137| Gene Symbol: Olfr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146488.1 | NP_666699.1 | 1 | 916 | ||
| GenBank mRNA | BC148231.1 | - | 1 | 920 | 40142242 | |