siRNA Details
Gene Symbol: Olfr722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146494.2 | NP_666705.1 | 2 | 1414 | ||
GenBank mRNA | BC051403.1 | - | 2 | 1414 | 6392119 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146494.2 | NP_666705.1 | 2 | 1414 | ||
GenBank mRNA | BC051403.1 | - | 2 | 1414 | 6392119 |