siRNA Details
Gene Symbol: Olfr1186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC132093.1 | - | 1 | 493 | 40130370 | |
BC132095.1 | - | 1 | 493 | 40130372 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC132093.1 | - | 1 | 493 | 40130370 | |
BC132095.1 | - | 1 | 493 | 40130372 |