siRNA Details
| Gene Symbol: LCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005564.4 | NP_005555.2 | 4 | 553 | ||
| GenBank mRNA | AK301694.1 | - | 4 | 530 | ||
| AK316217.1 | - | 4 | 530 | |||
| BC033089.1 | - | 4 | 517 | 5421124 | ||
| BP272828.1 | - | 4 | 530 | |||
| BX644845.1 | - | 4 | 566 | |||
| CA454137.1 | - | 4 | 515 | |||
| CR542092.1 | - | 4 | 457 | |||
| EU644752.1 | - | 4 | 463 | |||
| S75256.1 | - | 4 | 397 | |||
| X83006.1 | - | 4 | 520 | |||

LCN2
Genomic Map