siRNA Details
| Gene Symbol: Olfr1368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146534.1 | NP_666745.1 | 1 | 521 | ||
| GenBank mRNA | BC125382.1 | - | 1 | 547 | 40129897 | |
| BC125384.1 | - | 1 | 547 | 40129899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1368| Gene Symbol: Olfr1368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146534.1 | NP_666745.1 | 1 | 521 | ||
| GenBank mRNA | BC125382.1 | - | 1 | 547 | 40129897 | |
| BC125384.1 | - | 1 | 547 | 40129899 | ||