siRNA Details
Gene Symbol: Olfr1370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146535.1 | NP_666746.1 | 1 | 400 | ||
GenBank mRNA | BC110468.1 | - | 1 | 400 | 40043199 | |
BC110469.1 | - | 1 | 399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146535.1 | NP_666746.1 | 1 | 400 | ||
GenBank mRNA | BC110468.1 | - | 1 | 400 | 40043199 | |
BC110469.1 | - | 1 | 399 |