siRNA Details
| Gene Symbol: Olfr788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146551.1 | NP_666762.1 | 1 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr788| Gene Symbol: Olfr788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146551.1 | NP_666762.1 | 1 | 497 | ||