siRNA Details
| Gene Symbol: Olfr843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146567.2 | NP_666778.2 | 1 | 968 | ||
| GenBank mRNA | BC138386.1 | - | 1 | 968 | 8861406 | |
| BC138387.1 | - | 1 | 968 | 8861407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr843| Gene Symbol: Olfr843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146567.2 | NP_666778.2 | 1 | 968 | ||
| GenBank mRNA | BC138386.1 | - | 1 | 968 | 8861406 | |
| BC138387.1 | - | 1 | 968 | 8861407 | ||