siRNA Details
| Gene Symbol: Olfr700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146600.1 | NP_666811.1 | 1 | 392 | ||
| GenBank mRNA | BC104090.1 | - | 1 | 392 | ||
| BC104091.2 | - | 1 | 392 | 40042950 | ||
| BC106811.1 | - | 1 | 392 | |||
| BC106812.2 | - | 1 | 392 | 40043224 | ||

Olfr700
Genomic Map