siRNA Details
| Gene Symbol: Olfr303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146619.1 | NP_666830.1 | 1 | 798 | ||
| GenBank mRNA | BC132005.1 | - | 1 | 837 | 40130282 | |
| BC145991.1 | - | 1 | 837 | 40131294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr303| Gene Symbol: Olfr303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146619.1 | NP_666830.1 | 1 | 798 | ||
| GenBank mRNA | BC132005.1 | - | 1 | 837 | 40130282 | |
| BC145991.1 | - | 1 | 837 | 40131294 | ||