siRNA Details
| Gene Symbol: Olfr123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146630.1 | NP_666841.1 | 1 | 480 | ||
| GenBank mRNA | BC148279.1 | - | 1 | 480 | 40136837 | |
| BC148324.1 | - | 1 | 480 | 40136838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr123| Gene Symbol: Olfr123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146630.1 | NP_666841.1 | 1 | 480 | ||
| GenBank mRNA | BC148279.1 | - | 1 | 480 | 40136837 | |
| BC148324.1 | - | 1 | 480 | 40136838 | ||