siRNA Details
| Gene Symbol: Olfr513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146723.1 | NP_666934.1 | 1 | 789 | ||
| GenBank mRNA | BC107332.1 | - | 1 | 789 | 40054956 | |
| BC107333.1 | - | 1 | 789 | 40054960 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr513| Gene Symbol: Olfr513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146723.1 | NP_666934.1 | 1 | 789 | ||
| GenBank mRNA | BC107332.1 | - | 1 | 789 | 40054956 | |
| BC107333.1 | - | 1 | 789 | 40054960 | ||