siRNA Details
Gene Symbol: Olfr1362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146744.2 | NP_666955.2 | 1 | 176 | ||
GenBank mRNA | BC119336.1 | - | 1 | 176 | 8734085 | |
BC119338.1 | - | 1 | 176 | 8734087 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146744.2 | NP_666955.2 | 1 | 176 | ||
GenBank mRNA | BC119336.1 | - | 1 | 176 | 8734085 | |
BC119338.1 | - | 1 | 176 | 8734087 |