siRNA Details
| Gene Symbol: Olfr689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146750.1 | NP_666961.1 | 1 | 179 | ||
| GenBank mRNA | BC119379.1 | - | 1 | 203 | 8734128 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr689| Gene Symbol: Olfr689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146750.1 | NP_666961.1 | 1 | 179 | ||
| GenBank mRNA | BC119379.1 | - | 1 | 203 | 8734128 | |