siRNA Details
| Gene Symbol: Olfr414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146761.2 | NP_666972.2 | 1 | 590 | ||
| GenBank mRNA | BC106815.1 | - | 1 | 590 | 40043364 | |
| BC106816.1 | - | 1 | 590 | 40043365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr414| Gene Symbol: Olfr414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146761.2 | NP_666972.2 | 1 | 590 | ||
| GenBank mRNA | BC106815.1 | - | 1 | 590 | 40043364 | |
| BC106816.1 | - | 1 | 590 | 40043365 | ||