siRNA Details
Gene Symbol: Olfr1502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146797.1 | NP_667008.1 | 1 | 450 | ||
GenBank mRNA | BC106810.1 | - | 1 | 450 | 40042985 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146797.1 | NP_667008.1 | 1 | 450 | ||
GenBank mRNA | BC106810.1 | - | 1 | 450 | 40042985 |