siRNA Details
| Gene Symbol: Olfr143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146806.2 | NP_667017.2 | 1 | 605 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr143| Gene Symbol: Olfr143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146806.2 | NP_667017.2 | 1 | 605 | ||