siRNA Details
| Gene Symbol: Olfr923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146816.2 | NP_667027.2 | 1 | 620 | ||
| GenBank mRNA | BC132247.1 | - | 1 | 620 | 40130524 | |
| BC132249.1 | - | 1 | 620 | 40130526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr923| Gene Symbol: Olfr923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146816.2 | NP_667027.2 | 1 | 620 | ||
| GenBank mRNA | BC132247.1 | - | 1 | 620 | 40130524 | |
| BC132249.1 | - | 1 | 620 | 40130526 | ||