siRNA Details
| Gene Symbol: Olfr272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146839.1 | NP_667050.1 | 1 | 82 | ||
| GenBank mRNA | BC128006.1 | - | 1 | 86 | 40135971 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr272| Gene Symbol: Olfr272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146839.1 | NP_667050.1 | 1 | 82 | ||
| GenBank mRNA | BC128006.1 | - | 1 | 86 | 40135971 | |